siRNA Details
| Gene Symbol: C5ar2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001146005.1 | NP_001139477.1 | 2 | 2082 | ||
| NM_176912.4 | NP_795886.2 | 2 | 2175 | |||
| GenBank mRNA | AK053187.1 | - | 2 | 2176 | ||
| AK160050.1 | - | 2 | 2083 | |||
More Information
Set Membership: |
|

C5ar2
Genomic Map