siRNA Details
| Gene Symbol: Ctnna3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001164376.1 | NP_001157848.1 | 8 | 1231 | ||
| NM_001164519.1 | NP_001157991.1 | 3 | 390 | |||
| NM_177612.3 | NP_808280.2 | 8 | 1159 | |||
| GenBank mRNA | AF344871.1 | - | 8 | 1231 | ||
| AK030166.1 | - | 8 | 1160 | |||
| AK142745.1 | - | 8 | 1232 | |||
| BC112409.1 | - | 8 | 1133 | 40060582 | ||
| DQ380429.1 | - | 8 | 1159 | |||
| DQ380430.1 | - | 3 | 389 | |||

Ctnna3
Genomic Map