siRNA Details
| Gene Symbol: C130079G13Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177661.3 | NP_808329.1 | 5 | 915 | ||
| GenBank mRNA | AK081819.1 | - | 5 | 916 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

C130079G13Rik| Gene Symbol: C130079G13Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177661.3 | NP_808329.1 | 5 | 915 | ||
| GenBank mRNA | AK081819.1 | - | 5 | 916 | ||