siRNA Details
| Gene Symbol: Rgma | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177740.5 | NP_808408.2 | 4 | 2617 | ||
| GenBank mRNA | AJ557513.1 | - | 3 | 2371 | ||
| AK082560.1 | - | 3 | 2516 | |||
| BC023870.1 | - | 3 | 2421 | 5353958 | ||
| BC059072.1 | - | 4 | 2617 | 6826181 | ||

Rgma
Genomic Map