siRNA Details
Gene Symbol: Triml1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177742.4 | NP_808410.1 | 3 | 648 | ||
GenBank mRNA | AK077092.1 | - | 3 | 650 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Triml1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177742.4 | NP_808410.1 | 3 | 648 | ||
GenBank mRNA | AK077092.1 | - | 3 | 650 |