siRNA Details
| Gene Symbol: Esyt3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177775.3 | NP_808443.2 | 15, 16 | 1651 | ||
| GenBank mRNA | AK086367.1 | - | 15, 16 | 1652 | ||
| AK220532.1 | - | 15, 16 | 1639 | |||
| BC118967.2 | - | 1599 | 40097262 | |||

Esyt3
Genomic Map