siRNA Details
| Gene Symbol: Zfp786 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177882.4 | NP_808550.1 | 2 | 219 | ||
| GenBank mRNA | AK080438.1 | - | 2 | 219 | ||
| BC138897.1 | - | 2 | 160 | 8861919 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zfp786| Gene Symbol: Zfp786 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177882.4 | NP_808550.1 | 2 | 219 | ||
| GenBank mRNA | AK080438.1 | - | 2 | 219 | ||
| BC138897.1 | - | 2 | 160 | 8861919 | ||