siRNA Details
| Gene Symbol: Mob3b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178061.5 | NP_835162.1 | 4 | 1518 | ||
| GenBank mRNA | AK039841.1 | - | 4 | 1364 | ||
| BC020028.1 | - | 4 | 1499 | 4007373 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mob3b| Gene Symbol: Mob3b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178061.5 | NP_835162.1 | 4 | 1518 | ||
| GenBank mRNA | AK039841.1 | - | 4 | 1364 | ||
| BC020028.1 | - | 4 | 1499 | 4007373 | ||