siRNA Details
| Gene Symbol: KRT7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_005556.3 | NP_005547.3 | 4 | 777 | ||
| XM_011538325.1 | XP_011536627.1 | 4 | 730 | |||
| XM_017019294.1 | XP_016874783.1 | 4 | 503 | |||
| GenBank mRNA | AF509887.1 | - | 4 | 706 | ||
| AJ238246.1 | - | 4 | 711 | |||
| AK025881.1 | - | 4 | 704 | |||
| AK128505.1 | - | 4 | 596 | |||
| AK128848.1 | - | 5 | 632 | |||
| AK129924.1 | - | 5 | 711 | |||
| AK225498.1 | - | 4 | 700 | |||
| BC002700.2 | - | 4 | 692 | 3610347 | ||
| BC107082.1 | - | 1 | 36 | |||
| BE744321.1 | - | 354 | ||||
| X03212.1 | - | 4 | 711 | |||
| Z36852.1 | - | 2 | 117 | |||

KRT7
Genomic Map