siRNA Details
| Gene Symbol: Commd8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178599.4 | NP_848714.1 | 2 | 237 | ||
| GenBank mRNA | AK009275.1 | - | 2 | 226 | ||
| AK012653.1 | - | 2 | 223 | |||
| AK013015.1 | - | 2 | 208 | |||
| AK013371.1 | - | 2 | 210 | |||
| AK017777.1 | - | 2 | 152 | |||
| AK077934.1 | - | 2 | 237 | |||
| AK151560.1 | - | 2 | 220 | |||
| AK158078.1 | - | 2 | 220 | |||
| AK170011.1 | - | 2 | 210 | |||
| BC098183.1 | - | 2 | 213 | 5150466 | ||

Commd8
Genomic Map