siRNA Details
| Gene Symbol: Ubxn10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001285928.1 | NP_001272857.1 | 4 | 2602 | ||
| NM_001285929.1 | NP_001272858.1 | 3 | 2491 | |||
| NM_001285930.1 | NP_001272859.1 | 3 | 2355 | |||
| NM_178671.5 | NP_848786.1 | 2 | 2224 | |||
| GenBank mRNA | AK046949.1 | - | 4 | 2603 | ||
| AK077656.1 | - | 2 | 2227 | |||
| AK133185.1 | - | 2 | 2210 | |||
| AK161245.1 | - | 2 | 2211 | |||
| AK161296.1 | - | 2 | 2187 | |||
| BC027099.1 | - | 3 | 2353 | 5041000 | ||

Ubxn10
Genomic Map