siRNA Details
| Gene Symbol: Zfp385b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001113399.1 | NP_001106870.1 | 8 | 1951 | ||
| NM_001113400.1 | NP_001106871.1 | 8 | 1973 | |||
| NM_178723.6 | NP_848838.2 | 10 | 2483 | |||
| GenBank mRNA | AK046798.1 | - | 10 | 2485 | ||
| AK050293.1 | - | 8 | 1974 | |||
| AK078551.1 | - | 8 | 1929 | |||
| AK079322.1 | - | 10 | 2369 | |||
| AK149352.1 | - | 10 | 2470 | |||

Zfp385b
Genomic Map