siRNA Details
| Gene Symbol: Gulo | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178747.3 | NP_848862.1 | 12 | 1709 | ||
| GenBank mRNA | AK077740.1 | - | 12 | 1713 | ||
| AK167460.1 | - | 12 | 1697 | |||
| BC019856.1 | - | 12 | 1655 | 5123684 | ||
| BC028822.1 | - | 12 | 1664 | 5097681 | ||
| BC028828.1 | - | 12 | 1667 | 5101228 | ||
| BC034835.1 | - | 11 | 1464 | |||
| BC035221.1 | - | 1 | 88 | |||

Gulo
Genomic Map