siRNA Details
| Gene Symbol: Ano4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001277188.1 | NP_001264117.1 | 4 | 696 | ||
| GenBank mRNA | AK039466.1 | - | 4 | 498 | ||
| AK039728.1 | - | 4 | 506 | |||
| AK078364.1 | - | 4 | 460 | |||
| BC065114.1 | - | 3 | 591 | |||
| CF535597.1 | - | 3 | 591 | |||

Ano4
Genomic Map