siRNA Details
| Gene Symbol: Adck2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178873.3 | NP_849204.1 | 1 | 975 | ||
| GenBank mRNA | AK028557.1 | - | 1 | 979 | ||
| AK028568.1 | - | 1 | 976 | |||
| AK161225.1 | - | 1 | 978 | |||
| BC069944.1 | - | 1 | 723 | 30446117 | ||

Adck2
Genomic Map