siRNA Details
| Gene Symbol: Garnl3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178888.4 | NP_849219.2 | 5, 6 | 496 | ||
| GenBank mRNA | AK034068.1 | - | 5, 6 | 498 | ||
| AK036258.1 | - | 4, 5 | 792 | |||
| AK133163.1 | - | 6, 7 | 670 | |||
| BC057383.1 | - | 4, 5 | 490 | |||
| BC132143.1 | - | 5, 6 | 439 | 40130420 | ||

Garnl3
Genomic Map