siRNA Details
| Gene Symbol: LCP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_005565.3 | NP_005556.1 | 2 | 317 | ||
| GenBank mRNA | AK292890.1 | - | 2 | 278 | ||
| AK300000.1 | - | 2 | 296 | |||
| AK310369.1 | - | 2 | 258 | |||
| BC016618.1 | - | 2 | 254 | 4476274 | ||
| BG911397.1 | - | 2 | 317 | |||
| BT007273.1 | - | 2 | 110 | |||
| FN432830.1 | - | 2 | 110 | |||
| U20158.1 | - | 2 | 365 | |||

LCP2
Genomic Map