siRNA Details
| Gene Symbol: Rnpepl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_181405.4 | NP_852070.3 | 4 | 1043 | ||
| GenBank mRNA | AK154881.1 | - | 4 | 1044 | ||
| AK183521.1 | - | 2 | 125 | |||
| BC031789.1 | - | 4 | 463 | 4006916 | ||
| BQ559332.1 | - | 2 | 227 | |||
| CA315475.1 | - | 4 | 737 | |||

Rnpepl1
Genomic Map