siRNA Details
| Gene Symbol: Emilin3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001291145.1 | NP_001278074.1 | 2 | 403 | ||
| NM_182840.2 | NP_878260.1 | 2 | 403 | |||
| GenBank mRNA | BC002161.1 | - | 1 | 31 | ||
| BC054738.1 | - | 2 | 392 | 6402350 | ||
| BC057619.1 | - | 2 | 392 | 6402350 | ||

Emilin3
Genomic Map