siRNA Details
| Gene Symbol: Plk5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_183152.3 | NP_898975.2 | 8, 9 | 1091 | ||
| GenBank mRNA | AK031968.1 | - | 8, 9 | 1092 | ||
| BC086684.1 | - | 3, 4 | 388 | 30679265 | ||
| BC099679.1 | - | 8, 9 | 1060 | 6489647 | ||

Plk5
Genomic Map