siRNA Details
| Gene Symbol: Sowaha | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_183173.2 | NP_898996.2 | 1 | 2142 | ||
| GenBank mRNA | AK043538.1 | - | 1 | 2029 | ||
| AY496686.1 | - | 1 | 611 | |||
| BC117862.1 | - | 2 | 820 | |||
| BC117863.1 | - | 1 | 1480 | 40095300 | ||

Sowaha
Genomic Map