siRNA Details
| Gene Symbol: Nlrp9b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_194058.2 | NP_918947.2 | 2 | 234 | ||
| GenBank mRNA | AY360471.1 | - | 101 | |||
| AY596196.1 | - | 2 | 234 | |||
| BC141249.1 | - | 2 | 232 | 9055861 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Nlrp9b| Gene Symbol: Nlrp9b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_194058.2 | NP_918947.2 | 2 | 234 | ||
| GenBank mRNA | AY360471.1 | - | 101 | |||
| AY596196.1 | - | 2 | 234 | |||
| BC141249.1 | - | 2 | 232 | 9055861 | ||