siRNA Details
| Gene Symbol: Trim45 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001165953.1 | NP_001159425.1 | 7 | 2261 | ||
| NM_194343.2 | NP_919324.2 | 6 | 2208 | |||
| GenBank mRNA | AK076600.1 | - | 7 | 1837 | ||
| BC057358.1 | - | 6 | 2086 | 6835531 | ||

Trim45
Genomic Map