siRNA Details
| Gene Symbol: Morc2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001159288.1 | NP_001152760.1 | 8 | 1279 | ||
| NM_198162.1 | NP_937805.1 | 7 | 606 | |||
| GenBank mRNA | AK173042.1 | - | 8 | 1279 | ||
| BC059243.1 | - | 7 | 606 | 6417551 | ||

Morc2a
Genomic Map