siRNA Details
| Gene Symbol: Szt2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198170.4 | NP_937813.3 | 58 | 8059 | ||
| GenBank mRNA | AK032181.1 | - | 6 | 579 | ||
| AK129152.1 | - | 20 | 3491 | |||
| FJ998170.1 | - | 58 | 8059 | |||
| GU433214.1 | - | 58 | 7982 | |||

Szt2
Genomic Map