siRNA Details
| Gene Symbol: Szt2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198170.4 | NP_937813.3 | 69 | 9666 | ||
| GenBank mRNA | AK032181.1 | - | 18 | 2367 | ||
| BC059842.1 | - | 12 | 1572 | |||
| FJ998170.1 | - | 69 | 9666 | |||
| GU433214.1 | - | 69 | 9589 | |||

Szt2
Genomic Map