siRNA Details
| Gene Symbol: Lrrc4b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198250.1 | NP_937893.1 | 3 | 1783 | ||
| GenBank mRNA | AK043555.1 | - | 1 | 573 | ||
| AK140418.1 | - | 2 | 1730 | |||
| BC053486.1 | - | 2 | 1647 | |||
| BC060263.1 | - | 3 | 1783 | 5686281 | ||

Lrrc4b
Genomic Map