siRNA Details
| Gene Symbol: Them6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198607.1 | NP_941009.1 | 2 | 993 | ||
| GenBank mRNA | AK019809.1 | - | 2 | 605 | ||
| BC047266.1 | - | 2 | 993 | 6307114 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Them6| Gene Symbol: Them6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198607.1 | NP_941009.1 | 2 | 993 | ||
| GenBank mRNA | AK019809.1 | - | 2 | 605 | ||
| BC047266.1 | - | 2 | 993 | 6307114 | ||