siRNA Details
| Gene Symbol: 5031414D18Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198642.2 | NP_941044.3 | 10 | 1679 | ||
| GenBank mRNA | AK030303.1 | - | 5 | 2791 | ||
| AK155201.1 | - | 10 | 1665 | |||
| AK170426.1 | - | 10 | 1643 | |||
| BC052668.1 | - | 10 | 1691 | 30054870 | ||

5031414D18Rik
Genomic Map