siRNA Details
| Gene Symbol: Ccdc58 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001159421.1 | NP_001152893.1 | 4 | 877 | ||
| NM_001159422.1 | NP_001152894.1 | 5 | 654 | |||
| NM_198645.2 | NP_941047.1 | 5 | 1003 | |||
| GenBank mRNA | AK135450.1 | - | 5 | 583 | ||
| BB686073.1 | - | 3 | 318 | |||
| BC024406.1 | - | 5 | 567 | |||
| BC024862.1 | - | 5 | 979 | 5356370 | ||
| BC053690.1 | - | 5 | 555 | |||

Ccdc58
Genomic Map