siRNA Details
| Gene Symbol: Tbc1d2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198664.3 | NP_941066.3 | 1 | 295 | ||
| GenBank mRNA | AK041359.1 | - | 1 | 307 | ||
| AK148254.1 | - | 1 | 298 | |||
| AK148289.1 | - | 1 | 212 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tbc1d2| Gene Symbol: Tbc1d2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198664.3 | NP_941066.3 | 1 | 295 | ||
| GenBank mRNA | AK041359.1 | - | 1 | 307 | ||
| AK148254.1 | - | 1 | 298 | |||
| AK148289.1 | - | 1 | 212 | |||