siRNA Details
| Gene Symbol: MAGEC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_005462.4 | NP_005453.2 | 4 | 3577 | ||
| XM_011531418.2 | XP_011529720.1 | 4 | 3513 | |||
| GenBank mRNA | AF056334.1 | - | 1 | 3539 | ||
| BC127771.1 | - | 3 | 827 | |||
| BC153867.1 | - | 3579 | ||||

MAGEC1
Genomic Map