siRNA Details
| Gene Symbol: 4930590J08Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198668.2 | NP_941070.1 | 8 | 1377 | ||
| GenBank mRNA | AK029822.1 | - | 8 | 1377 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

4930590J08Rik| Gene Symbol: 4930590J08Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198668.2 | NP_941070.1 | 8 | 1377 | ||
| GenBank mRNA | AK029822.1 | - | 8 | 1377 | ||