siRNA Details
| Gene Symbol: Lkaaear1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_199023.3 | NP_950188.2 | 4 | 594 | ||
| GenBank mRNA | AK076877.1 | - | 4 | 595 | ||
| BC087873.1 | - | 4 | 582 | 6740628 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Lkaaear1| Gene Symbol: Lkaaear1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_199023.3 | NP_950188.2 | 4 | 594 | ||
| GenBank mRNA | AK076877.1 | - | 4 | 595 | ||
| BC087873.1 | - | 4 | 582 | 6740628 | ||