siRNA Details
| Gene Symbol: 4922502D21Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_199034.3 | NP_950199.1 | 5 | 1350 | ||
| GenBank mRNA | AK076635.1 | - | 5 | 1323 | ||
| AK132711.1 | - | 5 | 1335 | |||
| BC089479.1 | - | 5 | 1350 | 6704696 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

4922502D21Rik| Gene Symbol: 4922502D21Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_199034.3 | NP_950199.1 | 5 | 1350 | ||
| GenBank mRNA | AK076635.1 | - | 5 | 1323 | ||
| AK132711.1 | - | 5 | 1335 | |||
| BC089479.1 | - | 5 | 1350 | 6704696 | ||