siRNA Details
| Gene Symbol: Lrsam1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_199302.2 | NP_955006.1 | 18, 19 | 1664 | ||
| GenBank mRNA | AK211988.1 | - | 2, 3 | 149 | ||
| BC049146.1 | - | 18, 19 | 1660 | 6306042 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Lrsam1| Gene Symbol: Lrsam1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_199302.2 | NP_955006.1 | 18, 19 | 1664 | ||
| GenBank mRNA | AK211988.1 | - | 2, 3 | 149 | ||
| BC049146.1 | - | 18, 19 | 1660 | 6306042 | ||