siRNA Details
| Gene Symbol: Lrsam1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_199302.2 | NP_955006.1 | 25 | 2721 | ||
| GenBank mRNA | BC049146.1 | - | 25 | 2717 | 6306042 | |
| Gene Symbol: 4930555K19Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK016133.1 | - | 472 | |||

Lrsam1
Genomic Map