siRNA Details
| Gene Symbol: Eml4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001114361.1 | NP_001107833.1 | 8 | 1022 | ||
| NM_001114362.1 | NP_001107834.1 | 6 | 815 | |||
| NM_001286567.1 | NP_001273496.1 | 8 | 925 | |||
| NM_199466.3 | NP_955760.3 | 5 | 686 | |||
| GenBank mRNA | AK144604.1 | - | 8 | 1023 | ||
| BC065178.1 | - | 8 | 863 | 6827397 | ||
| BC066099.1 | - | 5 | 617 | 30533573 | ||
| BC067011.1 | - | 5 | 635 | 6405184 | ||
| BC070427.1 | - | 6 | 755 | 6413041 | ||
| BC079619.1 | - | 5 | 617 | 30533573 | ||
| DQ230326.1 | - | 7 | 705 | |||

Eml4
Genomic Map