siRNA Details
| Gene Symbol: Zcchc5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_199468.1 | NP_955762.1 | 2 | 2789 | ||
| GenBank mRNA | AK084983.1 | - | 2 | 2696 | ||
| BC064821.1 | - | 2 | 2789 | 30139133 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zcchc5| Gene Symbol: Zcchc5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_199468.1 | NP_955762.1 | 2 | 2789 | ||
| GenBank mRNA | AK084983.1 | - | 2 | 2696 | ||
| BC064821.1 | - | 2 | 2789 | 30139133 | ||