siRNA Details
| Gene Symbol: Nploc4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001195023.1 | NP_001181952.1 | 4 | 448 | ||
| NM_199469.2 | NP_955763.1 | 4 | 448 | |||
| GenBank mRNA | AK129375.1 | - | 4 | 423 | ||
| AK141998.1 | - | 4 | 447 | |||
| AK149823.1 | - | 4 | 420 | |||
| AK149912.1 | - | 4 | 447 | |||
| BC065156.1 | - | 4 | 416 | 5706754 | ||

Nploc4
Genomic Map