siRNA Details
| Gene Symbol: INSL5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_005478.4 | NP_005469.2 | 2 | 588 | ||
| GenBank mRNA | AF133816.2 | - | 2 | 596 | ||
| AY359030.1 | - | 2 | 588 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

INSL5| Gene Symbol: INSL5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_005478.4 | NP_005469.2 | 2 | 588 | ||
| GenBank mRNA | AF133816.2 | - | 2 | 596 | ||
| AY359030.1 | - | 2 | 588 | |||