siRNA Details
| Gene Symbol: BC055324 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_201364.1 | NP_958752.1 | 8, 9 | 942 | ||
| GenBank mRNA | AK031594.1 | - | 8, 9 | 938 | ||
| AK141661.1 | - | 8, 9 | 946 | |||
| AK163374.1 | - | 846 | ||||
| BC052855.1 | - | 8, 9 | 936 | 30058659 | ||
| BC055324.1 | - | 8, 9 | 942 | 5715852 | ||

BC055324
Genomic Map