siRNA Details
Gene Symbol: Gpr176 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_201367.3 | NP_958755.1 | 3 | 2768 | ||
GenBank mRNA | AK045979.1 | - | 1 | 646 | ||
BC052037.1 | - | 3 | 2660 | 5702597 |

More Information
Set Membership: |
![]() |
Panther Classification:
Gene Ontology Categories:
