siRNA Details
| Gene Symbol: Ccdc84 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_201372.3 | NP_958760.2 | 3 | 332 | ||
| GenBank mRNA | AK052757.1 | - | 3 | 347 | ||
| AK162606.1 | - | 3 | 365 | |||
| AK171754.1 | - | 339 | ||||
| BC055929.1 | - | 3 | 332 | |||
| BC096560.1 | - | 3 | 337 | 1527981 | ||
| BC116877.1 | - | 3 | 346 | 40126196 | ||
| BC116879.1 | - | 3 | 346 | 40126198 | ||
| BC144712.1 | - | 3 | 346 | |||
| BC144713.1 | - | 3 | 346 | |||

Ccdc84
Genomic Map