siRNA Details
| Gene Symbol: Ccdc84 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_201372.3 | NP_958760.2 | 7 | 682 | ||
| GenBank mRNA | AK052757.1 | - | 7 | 725 | ||
| AK162606.1 | - | 5 | 1098 | |||
| AK171754.1 | - | 1072 | ||||
| BC055929.1 | - | 7 | 682 | |||
| BC096560.1 | - | 7 | 687 | 1527981 | ||

Ccdc84
Genomic Map