siRNA Details
| Gene Symbol: PARP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001042618.1 | NP_001036083.1 | 8 | 714 | ||
| NM_005484.3 | NP_005475.2 | 8 | 753 | |||
| XM_005267247.3 | XP_005267304.1 | 8 | 753 | |||
| XM_017020912.1 | XP_016876401.1 | 8 | 1287 | |||
| GenBank mRNA | AF085734.1 | - | 8 | 750 | ||
| AJ236876.2 | - | 7 | 664 | |||
| AJ236912.1 | - | 8 | 688 | |||
| AK290079.1 | - | 8 | 693 | |||
| AK300971.1 | - | 6 | 471 | |||

PARP2
Genomic Map