siRNA Details
| Gene Symbol: Ccdc155 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC050196.1 | - | 3 | 506 | 30008752 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ccdc155| Gene Symbol: Ccdc155 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC050196.1 | - | 3 | 506 | 30008752 | |