siRNA Details
| Gene Symbol: Flrt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_201411.2 | NP_958813.1 | 2 | 2510 | ||
| GenBank mRNA | AK031503.1 | - | 2 | 2510 | ||
| AY495667.1 | - | 1 | 596 | |||
| AY495668.1 | - | 2 | 2465 | |||
| BC070403.1 | - | 2 | 2514 | 5696478 | ||
| BC111879.1 | - | 2 | 2213 | 40059728 | ||
| BC112383.1 | - | 2 | 2213 | 40059727 | ||
| BC138214.1 | - | 2 | 2151 | 8861233 | ||

Flrt1
Genomic Map