siRNA Details
| Gene Symbol: H2-M10.3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AY211080.1 | - | 6 | 1180 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

H2-M10.3| Gene Symbol: H2-M10.3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AY211080.1 | - | 6 | 1180 | ||