siRNA Details
| Gene Symbol: Rwdd4a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_203507.3 | NP_987103.1 | 4 | 542 | ||
| GenBank mRNA | AK004597.1 | - | 4 | 496 | ||
| AK005797.1 | - | 4 | 527 | |||
| AK016505.1 | - | 4 | 516 | |||
| AK185756.1 | - | 3 | 195 | |||
| BC016198.1 | - | 4 | 478 | 4503404 | ||

Rwdd4a
Genomic Map